Assay Details
| Gene Symbol: serinc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213514.1 | NP_998679.1 | 11 - 12 | 1336 | 76 | |
| GenBank mRNA | BC044159.1 | - | 11 - 12 | 1336 | 76 | |
| BC071500.1 | - | 11 - 12 | 1369 | 76 | ||
| BC165736.1 | - | 11 - 12 | 1276 | 76 | ||
More Information
Set Membership: |
|

serinc5
Genomic Map