Assay Details
| Gene Symbol: etv5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126461.2 | NP_001119933.1 | 9 - 10 | 1263 | 64 | |
| NM_001328592.1 | NP_001315521.1 | 9 - 10 | 1263 | 64 | ||
| XM_021478612.1 | XP_021334287.1 | 9 - 10 | 1160 | 64 | ||
| XM_021478613.1 | XP_021334288.1 | 9 - 10 | 1255 | 64 | ||
| XM_021478614.1 | XP_021334289.1 | 9 - 10 | 1255 | 64 | ||
| GenBank mRNA | BC048059.1 | - | 9 - 10 | 1202 | 64 | |
| BC155729.1 | - | 9 - 10 | 1216 | 64 | ||
| BC165761.1 | - | 8 - 9 | 1025 | 64 | ||
More Information
Set Membership: |
|

etv5a
Genomic Map