Assay Details
| Gene Symbol: asf1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214757.2 | NP_999922.1 | 3 - 4 | 796 | 56 | |
| GenBank mRNA | BC066755.1 | - | 3 - 4 | 796 | 56 | |
More Information
Set Membership: |
|

asf1bb
Genomic Map