Assay Details
| Gene Symbol: ube2z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002330.5 | NP_001002330.2 | 7 | 1830 | 172 | |
| GenBank mRNA | BC076491.1 | - | 7 | 1446 | 174 | |
More Information
Set Membership: |
|

ube2z
Genomic Map