Assay Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081777.1 | NP_001075246.1 | 1 - 2 | 64 | 69 | |
| GenBank mRNA | AB292249.1 | - | 1 - 2 | 64 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001081777.1 transcripts.
Set Membership: |
|

FMOD
Genomic Map