Assay Details
| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081814.2 | NP_001075283.2 | 3 - 4 | 259 | 88 | |
| GenBank mRNA | AB025570.1 | - | 3 - 4 | 262 | 88 | |
| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628306.1 | XP_023484074.1 | 397 | 88 | ||
More Information
Set Membership: |
|

CHGA
Genomic Map