Assay Details
| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081814.2 | NP_001075283.2 | 4 - 5 | 329 | 75 | |
| GenBank mRNA | AB025570.1 | - | 4 - 5 | 332 | 75 | |
| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628306.1 | XP_023484074.1 | 467 | 75 | ||
More Information
Set Membership: |
|

CHGA
Genomic Map