Assay Details
| Gene Symbol: S100G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081890.1 | NP_001075359.1 | 1 - 2 | 138 | 75 | |
| XM_005613939.3 | XP_005613996.2 | 1212 | 75 | |||
| XM_005613940.3 | XP_005613997.2 | 1231 | 75 | |||
| GenBank mRNA | AY229893.1 | - | 1 - 2 | 138 | 75 | |
More Information
Set Membership: |
|

S100G
Genomic Map