Assay Details
| Gene Symbol: HSD3B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081911.1 | NP_001075380.1 | 1 - 2 | 260 | 59 | |
| NM_001308637.1 | NP_001295566.1 | 2 - 3 | 187 | 59 | ||
| GenBank mRNA | AF031665.1 | - | 2 - 3 | 187 | 59 | |
| D89666.1 | - | 1 - 2 | 260 | 59 | ||
More Information
Set Membership: |
|

HSD3B2
Genomic Map