Assay Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081934.2 | NP_001075403.1 | 1 - 2 | 702 | 77 | |
| XM_005599136.3 | XP_005599193.2 | 1191 | 77 | |||
| XM_005599137.3 | XP_005599194.2 | 1345 | 77 | |||
| XM_023616899.1 | XP_023472667.1 | 1098 | 77 | |||
| XM_023616900.1 | XP_023472668.1 | 1228 | 77 | |||
| XM_023616901.1 | XP_023472669.1 | 1261 | 77 | |||
| GenBank mRNA | AJ698508.1 | - | 1 - 2 | 702 | 77 | |
More Information
Set Membership: |
|

SLC26A2
Genomic Map