Assay Details
| Gene Symbol: KIR3DL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082518.2 | NP_001075987.2 | 1 - 2 | 76 | 84 | |
| GenBank mRNA | AB120394.1 | - | 1 - 2 | 77 | 84 | |
| AB120395.1 | - | 1 - 2 | 77 | 84 | ||
| KC412060.1 | - | 1 - 2 | 54 | 84 | ||
| Gene Symbol: KIR-ILTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB120397.1 | - | 1 - 2 | 57 | 84 | |
| AB120400.1 | - | 1 - 2 | 79 | 84 | ||
| AB120401.1 | - | 1 - 2 | 74 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001082518.2 transcripts.
Set Membership: |
|

KIR3DL
Genomic Map