Assay Details
| Gene Symbol: KIR3DL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082518.2 | NP_001075987.2 | 7 - 8 | 1159 | 59 | |
| GenBank mRNA | AB120394.1 | - | 7 - 8 | 1160 | 59 | |
| AB120395.1 | - | 7 - 8 | 1160 | 59 | ||
| KC412060.1 | - | 7 - 8 | 1137 | 59 | ||
More Information
Set Membership: |
|

KIR3DL
Genomic Map