Assay Details
| Gene Symbol: IL23A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082522.2 | NP_001075991.1 | 3 - 4 | 484 | 92 | |
| XM_014740754.2 | XP_014596240.1 | 1722 | 92 | |||
| GenBank mRNA | EU438773.1 | - | 3 - 4 | 484 | 92 | |
More Information
Set Membership: |
|

IL23A
Genomic Map