Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083951.2 | NP_001077420.2 | 2 - 3 | 293 | 95 | |
| GenBank mRNA | AF062377.1 | - | 2 - 3 | 201 | 95 | |
| AY184956.1 | - | 2 - 3 | 296 | 95 | ||
| JL626338.1 | - | 2 - 3 | 224 | 95 | ||
More Information
Set Membership: |
|

CXCL8
Genomic Map