Assay Details
| Gene Symbol: GCNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105534.3 | NP_001099004.2 | 1 - 2 | 72 | 122 | |
| GenBank mRNA | EU124678.1 | - | 1 - 2 | 92 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105534.3 transcripts.
Set Membership: |
|

GCNT1
Genomic Map