Assay Details
| Gene Symbol: CCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114941.1 | NP_001108413.1 | 2 - 3 | 207 | 87 | |
| XM_023651619.1 | XP_023507387.1 | 453 | 87 | |||
| GenBank mRNA | EU438775.1 | - | 2 - 3 | 207 | 87 | |
More Information
Set Membership: |
|

CCL3
Genomic Map