Assay Details
| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496235.5 | XP_001496285.2 | 494 | 67 | ||
| GenBank mRNA | AF076782.1 | - | 1 - 2 | 51 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDKN2B| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496235.5 | XP_001496285.2 | 494 | 67 | ||
| GenBank mRNA | AF076782.1 | - | 1 - 2 | 51 | 67 | |
Set Membership: |
|