Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163863.2 | NP_001157335.1 | 13 - 14 | 2076 | 71 | |
| XM_014732943.2 | XP_014588429.1 | 2009 | 71 | |||
| XM_014732944.2 | XP_014588430.1 | 2007 | 71 | |||
| XM_014732945.2 | XP_014588431.1 | 2004 | 71 | |||
| XM_014732946.2 | XP_014588432.1 | 2001 | 71 | |||
| XM_014732947.2 | XP_014588433.1 | 2010 | 71 | |||
| XM_014732950.2 | XP_014588436.1 | 1741 | 71 | |||
| XM_014732951.2 | XP_014588437.1 | 1738 | 71 | |||
| XM_014732952.2 | XP_014588438.1 | 1664 | 71 | |||
| XM_014732953.2 | XP_014588439.1 | 1661 | 71 | |||
| XM_014732954.2 | XP_014588440.1 | 1657 | 71 | |||
| XM_014732955.2 | XP_014588441.1 | 1656 | 71 | |||
| GenBank mRNA | AJ319907.1 | - | 1 - 2 | 43 | 71 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map