Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163863.2 | NP_001157335.1 | 14 - 15 | 2273 | 114 | |
| XM_014732943.2 | XP_014588429.1 | 2206 | 114 | |||
| XM_014732944.2 | XP_014588430.1 | 2204 | 114 | |||
| XM_014732945.2 | XP_014588431.1 | 2201 | 114 | |||
| XM_014732946.2 | XP_014588432.1 | 2198 | 114 | |||
| XM_014732947.2 | XP_014588433.1 | 2207 | 114 | |||
| XM_014732950.2 | XP_014588436.1 | 1938 | 114 | |||
| XM_014732951.2 | XP_014588437.1 | 1935 | 114 | |||
| XM_014732952.2 | XP_014588438.1 | 1861 | 114 | |||
| XM_014732953.2 | XP_014588439.1 | 1858 | 114 | |||
| XM_014732954.2 | XP_014588440.1 | 1854 | 114 | |||
| XM_014732955.2 | XP_014588441.1 | 1853 | 114 | |||
| GenBank mRNA | AJ319907.1 | - | 2 - 3 | 240 | 114 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map