Assay Details
| Gene Symbol: FGFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163863.2 | NP_001157335.1 | 15 - 16 | 2389 | 84 | |
| XM_014732943.2 | XP_014588429.1 | 2322 | 84 | |||
| XM_014732944.2 | XP_014588430.1 | 2320 | 84 | |||
| XM_014732945.2 | XP_014588431.1 | 2317 | 84 | |||
| XM_014732946.2 | XP_014588432.1 | 2314 | 84 | |||
| XM_014732947.2 | XP_014588433.1 | 2323 | 84 | |||
| XM_014732950.2 | XP_014588436.1 | 2054 | 84 | |||
| XM_014732951.2 | XP_014588437.1 | 2051 | 84 | |||
| XM_014732952.2 | XP_014588438.1 | 1977 | 84 | |||
| XM_014732953.2 | XP_014588439.1 | 1974 | 84 | |||
| XM_014732954.2 | XP_014588440.1 | 1970 | 84 | |||
| XM_014732955.2 | XP_014588441.1 | 1969 | 84 | |||
| GenBank mRNA | AJ319907.1 | - | 3 - 4 | 356 | 84 | |
More Information
Set Membership: |
|

FGFR2
Genomic Map