Assay Details
| Gene Symbol: IGFBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308604.1 | NP_001295533.1 | 2 - 3 | 513 | 72 | |
| GenBank mRNA | AY752935.1 | - | 1 - 2 | 131 | 72 | |
| CX592997.1 | - | 2 - 3 | 242 | 72 | ||
More Information
Set Membership: |
|

IGFBP6
Genomic Map