Assay Details
Gene Symbol: MIR29B-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032844.1 | 1 | 40 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CD34 molecule | CD34 | -79.5kb | Sense | Chr. 5 - 27408832 - 27431628 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-29c | MIR29C | eca-mir-29c | 476.0 | Sense | Chr. 5 - 27511696 - 27511783 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
