Assay Details
| Gene Symbol: LOC102151116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603672.2 | XP_005603729.2 | 1011 | 86 | ||
| XM_023624195.1 | XP_023479963.1 | 869 | 86 | |||
More Information
Set Membership: |
|

LOC102151116
Genomic Map