Assay Details
Gene Symbol: MIR130B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032877.1 | 1 | 36 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-301b | MIR301B | eca-mir-301b | -235.0 | Sense | Chr. 8 - 5383486 - 5383582 on NCBI Build EquCab3.0 |
Downstream | uncharacterized LOC102147531 | LOC102147531 | 2.3kb | Sense | Chr. 8 - 5386191 - 5397257 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
