Assay Details
Gene Symbol: MIR489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032827.1 | 1 | 44 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-653 | MIR653 | eca-mir-653 | -632.0 | Sense | Chr. 4 - 37174800 - 37174884 on NCBI Build EquCab3.0 |
Downstream | tissue factor pathway inhibitor 2 | TFPI2 | 364.9kb | Sense | Chr. 4 - 37540549 - 37545506 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
