Assay Details
| Gene Symbol: MIR24-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032867.1 | 1 | 34 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-27a | MIR27A | eca-mir-27a | -73.0 | Sense | Chr. 7 - 46233206 - 46233282 on NCBI Build EquCab3.0 |
| Downstream | microRNA mir-8992 | MIR8992 | eca-mir-8992 | 1.8kb | Sense | Chr. 7 - 46235200 - 46235302 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR24-1
Genomic Map