Assay Details
Gene Symbol: MIR193B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032928.1 | 1 | 39 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-365-2 | MIR365-2 | eca-mir-365-2 | -4.8kb | Sense | Chr. 13 - 31845857 - 31845943 on NCBI Build EquCab3.0 |
Downstream | myocardin related transcription factor B | MRTFB | MKL2 | 45.4kb | Sense | Chr. 13 - 31896222 - 32082450 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
