Assay Details
Gene Symbol: MIR9113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128143.2 | 1 | 58 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-9115 | MIR9115 | eca-mir-9115 | -4.8kb | Anti-sense | Chr. 12 - 24266983 - 24267125 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-9116 | MIR9116 | eca-mir-9116 | 862.0 | Anti-sense | Chr. 12 - 24272907 - 24273043 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
