Assay Details
| Gene Symbol: MIR3200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128022.1 | 1 | 93 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dual specificity protein phosphatase 18 | LOC111774601 | -54.7kb | Anti-sense | Chr. 8 - 8376531 - 8382612 on NCBI Build EquCab3.0 | |
| Downstream | uncharacterized protein C5orf52 | LOC100630784 | C8H5orf52 | 38.2kb | Sense | Chr. 8 - 8475684 - 8482623 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR3200
Genomic Map