Assay Details
| Gene Symbol: MIR9019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128032.1 | 1 | 70 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytohesin 4 | CYTH4 | -21.5kb | Anti-sense | Chr. 28 - 35632760 - 35662979 on NCBI Build EquCab3.0 | |
| Downstream | extracellular leucine rich repeat and fibronectin type III domain containing 2 | ELFN2 | 32.1kb | Sense | Chr. 28 - 35716677 - 35719133 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9019
Genomic Map