Assay Details
| Gene Symbol: MIR9091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128114.1 | 1 | 84 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 569 | ZNF569 | -23.5kb | Anti-sense | Chr. 10 - 8646200 - 8704933 on NCBI Build EquCab3.0 | |
| Downstream | zinc finger protein 527 | ZNF527 | 4.2kb | Sense | Chr. 10 - 8732754 - 8784815 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9091
Genomic Map