Assay Details
Gene Symbol: MIR9092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128115.1 | 1 | 90 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | phospholipase A2 inhibitor and LY6/PLAUR domain containing | PINLYP | -2.4kb | Anti-sense | Chr. 10 - 14739599 - 14744458 on NCBI Build EquCab3.0 | |
Downstream | zinc finger protein 576 | ZNF576 | 6.9kb | Anti-sense | Chr. 10 - 14753916 - 14758996 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
