Assay Details
| Gene Symbol: MIR9104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128131.1 | 1 | 79 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytoglobin | CYGB | -41.0kb | Sense | Chr. 11 - 5634154 - 5643786 on NCBI Build EquCab3.0 | |
| Downstream | aralkylamine N-acetyltransferase | AANAT | 4.5kb | Anti-sense | Chr. 11 - 5689430 - 5692104 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9104
Genomic Map