Assay Details
| Gene Symbol: MIR9141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128175.1 | 1 | 95 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB26, member RAS oncogene family |
RAB26 | -39.4kb | Anti-sense | Chr. 13 - 41862282 - 41868578 on NCBI Build EquCab3.0 | |
| Downstream | TSC complex subunit 2 | TSC2 | 19.1kb | Anti-sense | Chr. 13 - 41927187 - 41966103 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9141
Genomic Map