Assay Details
| Gene Symbol: MIR1193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033002.1 | 1 | 61 | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-494 | MIR494 | eca-mir-494 | -326.0 | Sense | Chr. 24 - 43340801 - 43340881 on NCBI Build EquCab3.0 |
| Downstream | microRNA mir-543 | MIR543 | eca-mir-543 | 1.6kb | Sense | Chr. 24 - 43342877 - 43342948 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR1193
Genomic Map