Assay Details
| Gene Symbol: PAQR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256979.1 | NP_001243908.1 | 1 | 1020 | 91 | |
| XM_005603515.3 | XP_005603572.1 | 2102 | 91 | |||
| XM_023624049.1 | XP_023479817.1 | 1173 | 91 | |||
| XM_023624050.1 | XP_023479818.1 | 1248 | 91 | |||
| XM_023624051.1 | XP_023479819.1 | 1179 | 91 | |||
| XM_023624052.1 | XP_023479820.1 | 1450 | 91 | |||
More Information
Set Membership: |
|

PAQR8
Genomic Map