Assay Details
| Gene Symbol: ACKR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257078.1 | NP_001244007.1 | 1 | 1019 | 62 | |
| XM_005610601.3 | XP_005610658.1 | 1197 | 62 | |||
| XM_005610602.3 | XP_005610659.1 | 1558 | 62 | |||
| XM_023642191.1 | XP_023497959.1 | 1617 | 62 | |||
More Information
Set Membership: |
|

ACKR3
Genomic Map