Assay Details
| Gene Symbol: SSTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257099.1 | NP_001244028.1 | 1 | 1016 | 90 | |
| XM_023638669.1 | XP_023494437.1 | 2372 | 90 | |||
| XM_023638714.1 | XP_023494482.1 | 1994 | 90 | |||
More Information
Set Membership: |
|

SSTR1
Genomic Map