Assay Details
| Gene Symbol: P2RY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257152.1 | NP_001244081.1 | 1 | 1024 | 104 | |
| XM_005611911.3 | XP_005611968.1 | 1521 | 104 | |||
| XM_005611914.3 | XP_005611971.1 | 1426 | 104 | |||
| XM_005611915.3 | XP_005611972.1 | 1339 | 104 | |||
| XM_014741499.2 | XP_014596985.1 | 1629 | 104 | |||
| XM_023644160.1 | XP_023499928.1 | 1216 | 104 | |||
| XM_023644161.1 | XP_023499929.1 | 1291 | 104 | |||
| XM_023644162.1 | XP_023499930.1 | 1409 | 104 | |||
| XM_023644163.1 | XP_023499931.1 | 1158 | 104 | |||
More Information
Set Membership: |
|

P2RY2
Genomic Map