Assay Details
| Gene Symbol: PABPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257185.1 | NP_001244114.1 | 1 | 1123 | 97 | |
| XM_005614279.3 | XP_005614336.1 | 1485 | 97 | |||
More Information
Set Membership: |
|

PABPC5
Genomic Map