Assay Details
| Gene Symbol: COX4I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270956.1 | NP_001257885.1 | 3 - 4 | 324 | 76 | |
| XM_005604417.3 | XP_005604474.1 | 399 | 76 | |||
More Information
Set Membership: |
|

COX4I2
Genomic Map