Assay Details
| Gene Symbol: CXCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278930.2 | NP_001265859.2 | 2 - 3 | 304 | 118 | |
| GenBank mRNA | HQ889900.1 | - | 2 - 3 | 288 | 118 | |
| HQ889980.1 | - | 2 - 3 | 242 | 118 | ||
| HQ890098.1 | - | 2 - 3 | 212 | 118 | ||
| JL616120.1 | - | 2 - 3 | 304 | 118 | ||
More Information
Set Membership: |
|

CXCL11
Genomic Map