Assay Details
| Gene Symbol: CXCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278930.2 | NP_001265859.2 | 3 - 4 | 377 | 94 | |
| GenBank mRNA | HQ889980.1 | - | 3 - 4 | 315 | 94 | |
| HQ890098.1 | - | 3 - 4 | 285 | 94 | ||
| JL616120.1 | - | 3 - 4 | 377 | 94 | ||
More Information
Set Membership: |
|

CXCL11
Genomic Map