Assay Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308949.1 | NP_001295878.1 | 3 - 4 | 595 | 87 | |
| GenBank mRNA | HQ889973.1 | - | 3 - 4 | 569 | 87 | |
| JL641320.1 | - | 1 - 2 | 96 | 87 | ||
More Information
Set Membership: |
|

CCDC59
Genomic Map