Assay Details
| Gene Symbol: SC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309338.1 | NP_001296267.1 | 4 - 5 | 553 | 86 | |
| GenBank mRNA | JL616286.1 | - | 4 - 5 | 486 | 86 | |
More Information
Set Membership: |
|

SC5D
Genomic Map