Assay Details
| Gene Symbol: EQMCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278933.1 | NP_001265862.1 | 5 - 6 | 1015 | 112 | |
| XM_023624017.1 | XP_023479785.1 | 1187 | 112 | |||
| GenBank mRNA | DQ145616.2 | - | 5 - 6 | 1015 | 112 | |
| DQ145617.1 | - | 4 - 5 | 716 | 112 | ||
| DQ145618.1 | - | 4 - 5 | 745 | 112 | ||
| X79894.1 | - | 938 | 112 | |||
More Information
Set Membership: |
|

EQMCE1
Genomic Map