Assay Details
| Gene Symbol: EQMCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278933.1 | NP_001265862.1 | 6 - 7 | 1052 | 95 | |
| XM_023624017.1 | XP_023479785.1 | 1224 | 95 | |||
| GenBank mRNA | DQ145616.2 | - | 6 - 7 | 1052 | 95 | |
More Information
Set Membership: |
|

EQMCE1
Genomic Map