Assay Details
| Gene Symbol: LOC100063463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601334.3 | XP_005601391.1 | 3542 | 81 | ||
| XM_023621811.1 | XP_023477579.1 | 3566 | 81 | |||
| XM_023621812.1 | XP_023477580.1 | 4553 | 81 | |||
| XM_023621813.1 | XP_023477581.1 | 385 | 81 | |||
More Information
Set Membership: |
|

LOC100063463
Genomic Map