Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639374.1 | XP_023495142.1 | 1820 | 72 | ||
| XM_023639375.1 | XP_023495143.1 | 1906 | 72 | |||
| XM_023639376.1 | XP_023495144.1 | 1737 | 72 | |||
| XM_023639377.1 | XP_023495145.1 | 773 | 72 | |||
| XM_023639378.1 | XP_023495146.1 | 687 | 72 | |||
More Information
Set Membership: |
|

CCDC126
Genomic Map