Assay Details
| Gene Symbol: SEPTIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618445.1 | XP_023474213.1 | 1685 | 74 | ||
| GenBank mRNA | JL626070.1 | - | 2 - 3 | 286 | 74 | |
More Information
Set Membership: |
|

SEPTIN10
Genomic Map